Learn More
Aviva Systems Biology Corp CKB Antibody - C-terminal region (ARP63753_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63753_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CKB. It was validated on Western Blot. . Alternate names: B-CK, CKBBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-075