Learn More
Sino Biological CKB / Creatine kinase B type Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 14415T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CKB / Creatine kinase B type (rh CKB / Creatine kinase B type; Catalog#14415-H07B; P12277; Pro2-Lys381). CKB / Creatine kinase B type specific IgG was purified by Human CKB / Creatine kinase B type affinity chromatography.
Catalog No. 50-224-8428