Learn More
Cayman Chemical CKI-7 (hydrochloride), Purity: ≥98%, CAS: 1177141-67-1, 10 mg
SDP
Catalog No. 502940861
Encompass_Preferred
A CK1 inhibitor (Ki = 8.5 μM); elective for CK1 over CK2, PKC, CaMKII, and cyclic AMP-dependent protein kinase (IC50s = 90, >1,000, 195, and 550 μM, respectively); inhibits phosphorylation of a peptide substrate by Leishmania CK1.2 at 10 μM
Catalog No. 50-294-0861 Supplier Cayman Chemical Supplier No. 2826810 MG
A CK1 inhibitor (Ki = 8.5 μM); elective for CK1 over CK2, PKC, CaMKII, and cyclic AMP-dependent protein kinase (IC50s = 90, >1,000, 195, and 550 μM, respectively); inhibits phosphorylation of a peptide substrate by Leishmania CK1.2 at 10 μM