Learn More
Foxx Life Sciences Plastic Erlenmeyer Flasks for Cell Culture and Fermentation, 2L, Autoclavable Polycarbonate (PC), 53mm (53B) VersaCap, Non-Sterile, Baffled Bottom
Supplier:  Foxx Life Sciences 2485231OEM
Encompass_Preferred
248-5231-OEM: Flask, 2L, PC, 53mm Cap, Baffle. Foxx Life Science EZclear Erlenmeyer Shaker Flasks are molded from virgin, optically clear, non-leaching polycarbonate resin. Our flasks retain clarity after gamma sterilization, and comply with stringent FDA and USP requirements. Sterile flasks are additionally certified as non-pyrogenic and DNase/RNase-free, making them ideal for cell culture, microbial culture, plants, media preparation, storage, and all other applications. They also fit all standard shaking incubator clamps and can be placed on any platform. Foxx Life Sciences EZclear Flasks are available with a flat or baffled base for increased oxygenation and mixing. Flasks are compatible with all 53mm VersaCap technology and adapter inserts, enabling easy customization of top connections. Vented VersaCap with a 0.2m hydrophobic membrane can be purchased with the unit, or separately. All Flasks and VersaCap are autoclavable at 15 PSI and 121C for 20 minutes.
Catalog No. 50-105-0028