Learn More
Cayman Chemical cKItCD117 GnK Isoform EX
SDP
Catalog No. 502947394
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged c-Kit GNNK−isoform extracellular domain expressed in HEK293 cells • Amino acids: 26-516 • MW: 56.7 kDa
Catalog No. 50-294-7394 Supplier Cayman Chemical Supplier No. 33980100 UG
Source: Active recombinant human C-terminal His-tagged c-Kit GNNK−isoform extracellular domain expressed in HEK293 cells • Amino acids: 26-516 • MW: 56.7 kDa