Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ckk-e12 | 1432494-65-9 | 98.0% | C60H120N4O6 | 10MG
SDP

Catalog No. 5000259618
Encompass_Preferred

CKK-E12 is an ionizable lipid employed to formulate lipid nanoparticles for delivery of RNA-based research agents; it aids encapsulation and endosomal release and has reported selectivity for liver parenchymal cells in vivo. Recommended handling includes dissolution in DMSO and defined low-temperature storage to preserve material integrity.

  • Ionizable lipid for lipid nanoparticle formulation.
  • Facilitates encapsulation and endosomal release of RNA.
  • Demonstrated selectivity for liver parenchymal cells in vivo.
  • High purity (98.0%).
  • Soluble in DMSO at 50 mg/mL; ultrasonic agitation recommended.
  • Storage guidance: pure form at -20°C for long-term stability; in solvent at -80°C for extended storage.

Catalog No. 50-002-59618 Supplier Medchemexpress LLC Supplier No. HY13478110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CKK-E12 is an ionizable lipid employed to formulate lipid nanoparticles for delivery of RNA-based research agents; it aids encapsulation and endosomal release and has reported selectivity for liver parenchymal cells in vivo. Recommended handling includes dissolution in DMSO and defined low-temperature storage to preserve material integrity.

  • Ionizable lipid for lipid nanoparticle formulation.
  • Facilitates encapsulation and endosomal release of RNA.
  • Demonstrated selectivity for liver parenchymal cells in vivo.
  • High purity (98.0%).
  • Soluble in DMSO at 50 mg/mL; ultrasonic agitation recommended.
  • Storage guidance: pure form at -20°C for long-term stability; in solvent at -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.