Learn More
Aviva Systems Biology Corp CKLF Antibody - N-terminal region (ARP47947_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47947_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CKLF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CKLF. Alternate names: C32, CKLF1, CKLF2, CKLF3, CKLF4, HSPC224, UCK-1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59686