Learn More
Aviva Systems Biology Corp CCR2 Antibody - middle region (ARP58409_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58409_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CKR-2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCR2. Alternate names: CC-CKR-2, CCR2A, CCR2B, CD192, CKR2, CKR2A, CKR2B, CMKBR2, MCP-1-RApplicationWB. Species reactivity: Human, Pig, Rabbit, Rat
Catalog No. 50-014-53767