Learn More
CPC Scientific H-Leu-Gln-Asn-Arg-Arg-Gly-Leu-Asp-Leu-Leu-Phe-Leu-Lys-Glu-Gly-Gly-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP012A
Encompass
CKS-17 is a heptadecapeptide isolated from a highly conserved region of the immunosuppressive retroviral envelope protein p15E that is also capable of inhibiting lymphocyte proliferation, B cell activation, natural killer cell cytotoxicity, and monocyte-mediated tumor cytotoxicity. It has been found that CKs-17 leads to the rise of intracellular cAMP levels that may affect several physiological cellular functions.SEQUENCE: H-Leu-Gln-Asn-Arg-Arg-Gly-Leu-Asp-Leu-Leu-Phe-Leu-Lys-Glu-Gly-Gly-Leu-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: LQNRRGLDLLFLKEGGLMOLECULAR FORMULA: C87H148N26O24MOLECULAR WEIGHT: 1942.29STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [99273-04-8]SYNONYMS: RESEARCH AREA: Antimicrobial, AntiviralREFERENCES:Nelson M, Nelson DS, Cianciolo GJ, Snyderman R. Cancer Immunol Immunother. 30(2), 113-118, (1989)R. A. J. Oostendorp et al., Eur. J. Immunol., 22, 1505 (1992)SKU(s): PKCP-012A
Catalog No. 50-211-0165