Learn More
Foxx Life Sciences Autofil Sterile Polystyrene (PS) Media Storage Bottles with SECUREgrasp GL45 Cap, 1000mL, 24/CS
GREENER_CHOICE
Catalog No. 501050039
Encompass_Preferred
1173-RLS: Media Bottle, 1000 mL, PS, Sterile. Foxx Life Sciences' Polystyrene Media Bottles have been designed specifically for Life Science researchers to store common solutions such as tissue culture media, serum, and buffers. Our bottles are designed to handle full vacuum, and can be used with the Autofil filtration system. Our Polysytrene bottles are used for general purpose cell culture aqueous solutions. Each Autofil bottle has features that improve the handling of the bottle, especially when the bottle is wet. The narrow bottle shoulder, combined with the molded finger grips, improves the ability to securely grip the bottle with gloved hands. Our patented, SECUREgrasp bottle cap enables easy handling and carrying of the bottle. The innovative cap also allows for easy uncapping, tightening, and bottle stacking and provides a large surface for bottle labeling. Foxx Life Sciences' Autofil is manufactured in ISO 13485 clean room in North America.
Catalog No. 50-105-0039 Supplier Foxx Life Sciences Supplier No. 1173RLS
1173-RLS: Media Bottle, 1000 mL, PS, Sterile. Foxx Life Sciences' Polystyrene Media Bottles have been designed specifically for Life Science researchers to store common solutions such as tissue culture media, serum, and buffers. Our bottles are designed to handle full vacuum, and can be used with the Autofil filtration system. Our Polysytrene bottles are used for general purpose cell culture aqueous solutions. Each Autofil bottle has features that improve the handling of the bottle, especially when the bottle is wet. The narrow bottle shoulder, combined with the molded finger grips, improves the ability to securely grip the bottle with gloved hands. Our patented, SECUREgrasp bottle cap enables easy handling and carrying of the bottle. The innovative cap also allows for easy uncapping, tightening, and bottle stacking and provides a large surface for bottle labeling. Foxx Life Sciences' Autofil is manufactured in ISO 13485 clean room in North America.