Learn More
Foxx Life Sciences Autofil Sterile Autoclavable Polycarbonate (PC) Media Storage Bottles with SECUREgrasp GL45 Cap, 250mL, 24/CS
GREENER_CHOICE
Catalog No. 501050045
Encompass_Preferred
1180-RLS: Media Bottle, 250 mL, PC, Sterile. Foxx Life Sciences' Polycarbonate Media Bottles have been designed specifically for Life Science researchers to store common solutions such as tissue culture media, serum, and buffers. Our bottles are designed to handle full vacuum, and can be used with the Autofil filtration system. Our PC bottles are highly transparent, shatterproof, and have good alcohol resistance. They are gamma-sterilized and rated to withstand five autoclave cycles (bottle only). The bottles are sterilized in the USA. Each Autofil bottle has features that improve the handling of the bottle, especially when the bottle is wet. The narrow bottle shoulder, combined with the molded finger grips, improves the ability to securely grip the bottle with gloved hands. Our patented, SECUREgrasp bottle cap enables easy handling and carrying. The innovative cap also allows for easy uncapping, tightening, and bottle stacking, and provides a large surface for labeling.
Catalog No. 50-105-0045 Supplier Foxx Life Sciences Supplier No. 1180RLS
1180-RLS: Media Bottle, 250 mL, PC, Sterile. Foxx Life Sciences' Polycarbonate Media Bottles have been designed specifically for Life Science researchers to store common solutions such as tissue culture media, serum, and buffers. Our bottles are designed to handle full vacuum, and can be used with the Autofil filtration system. Our PC bottles are highly transparent, shatterproof, and have good alcohol resistance. They are gamma-sterilized and rated to withstand five autoclave cycles (bottle only). The bottles are sterilized in the USA. Each Autofil bottle has features that improve the handling of the bottle, especially when the bottle is wet. The narrow bottle shoulder, combined with the molded finger grips, improves the ability to securely grip the bottle with gloved hands. Our patented, SECUREgrasp bottle cap enables easy handling and carrying. The innovative cap also allows for easy uncapping, tightening, and bottle stacking, and provides a large surface for labeling.