Learn More
Foxx Life Sciences Autoclavable Polypropylene (PP) Centrifuge Bottles, 250mL with Screw Cap
GREENER_CHOICE
Catalog No. 501050065
Encompass_Preferred
1801-RLS: Centrifuge Bottle, PP, 250mL, Screw Cap. Foxx Life Sciences Autofil Polypropylene Centrifuge Bottles are designed specifically to assist life science researchers in processing samples for cell suspensions, cell lysates, tissue homogenates, and organ homogenates. The Autofil Centrifuge Bottle is available in 250ml or 500ml, and fits a wide range of rotors. Material options include polycarbonate or polypropylene, with your choice of a traditional screw cap or a leak-proof sealing cap with silicone O-ring. Our centrifuge bottles feature thick walls that hold up to reuse, along with wide mouths that simplify filling and pouring liquids. Each Autofil Centrifuge Bottle also features large, clear-molded graduation markings and a frosted area for convenient labeling. Polypropylene Centrifuge bottles are autoclavable. *Available with a standard cap which can be used for low speed applications, or the seal cap for leak-free processing.
Catalog No. 50-105-0065 Supplier Foxx Life Sciences Supplier No. 1801RLS
1801-RLS: Centrifuge Bottle, PP, 250mL, Screw Cap. Foxx Life Sciences Autofil Polypropylene Centrifuge Bottles are designed specifically to assist life science researchers in processing samples for cell suspensions, cell lysates, tissue homogenates, and organ homogenates. The Autofil Centrifuge Bottle is available in 250ml or 500ml, and fits a wide range of rotors. Material options include polycarbonate or polypropylene, with your choice of a traditional screw cap or a leak-proof sealing cap with silicone O-ring. Our centrifuge bottles feature thick walls that hold up to reuse, along with wide mouths that simplify filling and pouring liquids. Each Autofil Centrifuge Bottle also features large, clear-molded graduation markings and a frosted area for convenient labeling. Polypropylene Centrifuge bottles are autoclavable. *Available with a standard cap which can be used for low speed applications, or the seal cap for leak-free processing.