Learn More
Foxx Life Sciences Autoclavable Polycarbonate Centrifuge Bottles, 500mL with Sealing Closure
GREENER_CHOICE
Catalog No. 501050072
Encompass_Preferred
1808-RLS: Centrifuge Bottle, PC, 500mL, Seal Cap. Foxx Life Sciences Autofil Polycarbonate Centrifuge Bottles are designed specifically to assist life science researchers in processing samples for cell suspensions, cell lysates, tissue homogenates, and organ homogenates. Autofil Centrifuge Bottles are available in 250ml or 500ml, and fit a wide range of rotors. Material options include polycarbonate or polypropylene, with your choice of a traditional screw cap or a leak-proof sealing closure with silicone O-ring. Our centrifuge bottles feature thick walls that hold up to reuse, along with wide mouths that simplify filling and pouring liquids. Each Autofil Centrifuge Bottle also features large, clear-molded graduation markings and a frosted area for convenient labeling. *Available with a standard cap which can be used for low speed applications, or the seal cap for leak-free processing. Bottles are autoclavable.
Catalog No. 50-105-0072 Supplier Foxx Life Sciences Supplier No. 1808RLS
1808-RLS: Centrifuge Bottle, PC, 500mL, Seal Cap. Foxx Life Sciences Autofil Polycarbonate Centrifuge Bottles are designed specifically to assist life science researchers in processing samples for cell suspensions, cell lysates, tissue homogenates, and organ homogenates. Autofil Centrifuge Bottles are available in 250ml or 500ml, and fit a wide range of rotors. Material options include polycarbonate or polypropylene, with your choice of a traditional screw cap or a leak-proof sealing closure with silicone O-ring. Our centrifuge bottles feature thick walls that hold up to reuse, along with wide mouths that simplify filling and pouring liquids. Each Autofil Centrifuge Bottle also features large, clear-molded graduation markings and a frosted area for convenient labeling. *Available with a standard cap which can be used for low speed applications, or the seal cap for leak-free processing. Bottles are autoclavable.