Learn More
Foxx Life Sciences Heavy Duty Autoclavable Polypropylene (PP) Vacuum Bottle, 2L, with 53mm (53B) Tangle Free VersaCap, Two 1/4 Inch Hose Barbs and Sealable Vent Port
Catalog No. 501050051
Encompass_Preferred
165-2102-OEM: HD Bottle, 2L, PP, 53mm Cap, 1/4" HB. Foxx Life Sciences Heavy Duty Vacuum Bottles combine maximum ease of use and versatility, with the highest level of strength and durability. Bottles have thick walls that can hold near full vacuum for 24 hours, making them excellent replacements for glass HPLC containers, and ideal for applications involving liquid aspiration, cell harvesting, and degassification. PETG and PC bottles can hold near half vacuum for 24 hours at 7.4 PSI/382.5 mmHg. All HD Vacuum Bottles are compatible with either 53mm or 83mm VersaCap technology and adapters, enabling easy customization of the bottle to fit your application. HD Vacuum Bottles are available in HDPE, PP, EZclear PETG, and PC. Bottle material and volume are molded into the side of the bottle, along with metric graduation marks certified accurate within 5%. PP and PC Heavy Duty Vacuum Bottles, as well as all caps and adapters, are autoclavable at 15 PSI and 121C for 20 minutes.
Catalog No. 50-105-0051 Supplier Foxx Life Sciences Supplier No. 1652102OEM
165-2102-OEM: HD Bottle, 2L, PP, 53mm Cap, 1/4" HB. Foxx Life Sciences Heavy Duty Vacuum Bottles combine maximum ease of use and versatility, with the highest level of strength and durability. Bottles have thick walls that can hold near full vacuum for 24 hours, making them excellent replacements for glass HPLC containers, and ideal for applications involving liquid aspiration, cell harvesting, and degassification. PETG and PC bottles can hold near half vacuum for 24 hours at 7.4 PSI/382.5 mmHg. All HD Vacuum Bottles are compatible with either 53mm or 83mm VersaCap technology and adapters, enabling easy customization of the bottle to fit your application. HD Vacuum Bottles are available in HDPE, PP, EZclear PETG, and PC. Bottle material and volume are molded into the side of the bottle, along with metric graduation marks certified accurate within 5%. PP and PC Heavy Duty Vacuum Bottles, as well as all caps and adapters, are autoclavable at 15 PSI and 121C for 20 minutes.