Learn More
Aviva Systems Biology Corp Cks1b Antibody - N-terminal region (ARP61441_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61441_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cks1b. It was validated on Western Blot. . Alternate names: 2410005G18Rik, 2610005D03Rik, AA407784, Cks1, sid1334ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-379