Learn More
Adam Equipment CKT 40M - Cruiser Approved Bench Checkweighing Scales - Capacity: 90lb / 40kg, Readability: 0.02lb / 0.01kg
Catalog No. NC3964938
Encompass_Preferred
With reliable performance and durable construction, the NTEP-approved CKT-M is unmatched in value for industrial weighing. Ideal for many applications, the CKT-M quickly accomplishes basic weighing, parts counting, and check weighing, which helps users meet company and regulatory guidelines for setting prices based on weight. Repetitive weighing tasks are fast, with a backlit display that changes colors to show if a sample is under, over, or within the preset limits. The compact design is ideal for limited workspaces or when multiple scales are not an option.
Catalog No. NC3964938 Supplier Adam Equipment Supplier No. CKT 40M
With reliable performance and durable construction, the NTEP-approved CKT-M is unmatched in value for industrial weighing. Ideal for many applications, the CKT-M quickly accomplishes basic weighing, parts counting, and check weighing, which helps users meet company and regulatory guidelines for setting prices based on weight. Repetitive weighing tasks are fast, with a backlit display that changes colors to show if a sample is under, over, or within the preset limits. The compact design is ideal for limited workspaces or when multiple scales are not an option.