Learn More
Foxx Life Sciences EZgrip 10L (2.5 Gal) HDPE Carboy with Leakproof Spigot, 83B VersaCap Closure, Large Pinched Handles
Supplier:  Foxx Life Sciences 1522211OEM
Encompass_Preferred
152-2211-OEM: Carboy, 10L, HDPE, 83mm Cap, with leak-proof Spigot. Foxx Life Sciences EZgrip Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the container and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric graduation marks which are certified to 5% accuracy. Recessed spigot location allows for Carboys to be stored on any surface without overhang, and the Foxx Life Sciences Spigot is 100% LEAKPROOF. EZfil wide-mouth design. Molded in graduation marks certified to 5% accuracy. VersaCap offers a wide variety of top connection options. 13L Max Capacity.
Catalog No. 50-386-931