Learn More
Foxx Life Sciences EZgrip 10L (2.5 Gal) Polypropylene (PP) Carboy with Leakproof Spigot, 83B VersaCap Closure, Large Pinched Handles
Catalog No. 501050085
Encompass_Preferred
155-2211-OEM: Carboy, 10L, PP, 83mm Cap, with Spigot. Foxx Life Sciences EZgrip Polypropylene Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the carboy and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric graduation marks which are certified to 5% accuracy. Recessed spigot location allows for Carboys to be stored on any surface without overhang, and the Foxx Life Sciences Spigot is 100% LEAKPROOF. EZfil wide-mouth design. Molded in graduation marks certified to 5% accuracy. VersaCap offers a wide variety of top connection options. 13L Max Capacity. Autoclavable.
Catalog No. 50-105-0085 Supplier Foxx Life Sciences Supplier No. 1552211OEM
155-2211-OEM: Carboy, 10L, PP, 83mm Cap, with Spigot. Foxx Life Sciences EZgrip Polypropylene Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the carboy and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric graduation marks which are certified to 5% accuracy. Recessed spigot location allows for Carboys to be stored on any surface without overhang, and the Foxx Life Sciences Spigot is 100% LEAKPROOF. EZfil wide-mouth design. Molded in graduation marks certified to 5% accuracy. VersaCap offers a wide variety of top connection options. 13L Max Capacity. Autoclavable.