Learn More
Foxx Life Sciences EZgrip 20L (5 Gal) Polypropylene (PP) Carboy with Leakproof Spigot, Wide Mouth 120mm VersaCap Closure, Large Pinched Handles
Supplier:  Foxx Life Sciences 1553221OEM
Encompass_Preferred
155-3221-OEM: Carboy, 20L, PP, 120mm Cap, with Spigot. Foxx Life Sciences EZgrip Polypropylene Wide-Mouth Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the carboy and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric graduation marks which are certified to 5% accuracy. Recessed spigot location allows for Carboys to be stored on any surface without overhang, and the Foxx Life Sciences Spigot is 100% LEAKPROOF. EZfil wide-mouth design. VersaCap offers a wide variety of top connection options. 26.5L Max Capacity. Autoclavable.
Catalog No. 50-105-0089