Learn More
Foxx Life Sciences EZgrip 10L (2.5 Gal) Light Blocking Amber HDPE Space Saving Carboy with Leakproof Spigot, 83mm (83B) VersaCap and 13L Max Capacity, Large Pinched Handles
Catalog No. 50386933
Encompass_Preferred
15K-2211-OEM: Carboy, 10L, Amber HDPE, 83mm Cap, with Spigot. Foxx Life Sciences EZgrip Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the container and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric graduation marks which are certified to 5% accuracy. Recessed spigot location allows for Carboys to be stored on any surface without overhang, and the Foxx Life Sciences Spigot is 100% LEAKPROOF. 100% Leak-Proof Spigot available in certain models. Molded in graduation marks certified to 5% accuracy. VersaCap offers a wide variety of top connection options.
Catalog No. 50-386-933 Supplier Foxx Life Sciences Supplier No. 15K2211OEM
15K-2211-OEM: Carboy, 10L, Amber HDPE, 83mm Cap, with Spigot. Foxx Life Sciences EZgrip Carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use. The distinctive rectangular shape saves valuable bench space. Larger handles on top of the container and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric graduation marks which are certified to 5% accuracy. Recessed spigot location allows for Carboys to be stored on any surface without overhang, and the Foxx Life Sciences Spigot is 100% LEAKPROOF. 100% Leak-Proof Spigot available in certain models. Molded in graduation marks certified to 5% accuracy. VersaCap offers a wide variety of top connection options.