Learn More
Apexbio Technology LLC CL 316243 disodium salt 151126-84-0 10mg
SDP
Catalog No. 5000091731
Encompass_Preferred
CL 316243 disodium salt is a small-molecule agonist targeting the 3-adrenergic receptor It is designed to activate 3 receptors thereby modulating lipolytic and thermogenic pathways predominantly in adipose tissues CL 316243 disodium salt exerts its biological activity by selectively binding to 3-adrenergic receptors leading to enhanced adipocyte-mediated lipid metabolism In experimental rodent models administration of CL 316243 increases insulin sensitivity and reduces obesity-related hyperglycemia by promoting glucose uptake primarily in adipose tissue Based on these pharmacological properties CL 316243 disodium salt holds research potential in the study of 3 receptor-mediated metabolic mechanisms and the exploration of therapeutic strategies for metabolic disorders such as obesity and type 2 diabetes
Catalog No. 50-000-91731 Supplier Apexbio Technology LLC Supplier No. B676610
CL 316243 disodium salt is a small-molecule agonist targeting the 3-adrenergic receptor It is designed to activate 3 receptors thereby modulating lipolytic and thermogenic pathways predominantly in adipose tissues CL 316243 disodium salt exerts its biological activity by selectively binding to 3-adrenergic receptors leading to enhanced adipocyte-mediated lipid metabolism In experimental rodent models administration of CL 316243 increases insulin sensitivity and reduces obesity-related hyperglycemia by promoting glucose uptake primarily in adipose tissue Based on these pharmacological properties CL 316243 disodium salt holds research potential in the study of 3 receptor-mediated metabolic mechanisms and the exploration of therapeutic strategies for metabolic disorders such as obesity and type 2 diabetes