Learn More
Abcam CL-316243 disodium salt, adrenergic-beta3 receptor agonist, 25MG
Catalog No. 502583276
Encompass_Preferred
MW 465.8 Da, Purity >99%. Selective adrenergic-β3 receptor agonist. Induces adipose tissue remodelling in rat. Hypoglycemic agent. Active *in vivo* and *in vitro*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3276 Supplier Abcam Supplier No. AB14460525MG
MW 465.8 Da, Purity >99%. Selective adrenergic-β3 receptor agonist. Induces adipose tissue remodelling in rat. Hypoglycemic agent. Active *in vivo* and *in vitro*.
The product is subject to the following: Abcam Restricted Use Statement