Learn More
Abcam CL-316243 disodium salt, adrenergic-beta3 receptor agonist, 5MG
Catalog No. 502583275
Encompass_Preferred
MW 465.8 Da, Purity >99%. Selective adrenergic-β3 receptor agonist. Induces adipose tissue remodelling in rat. Hypoglycemic agent. Active *in vivo* and *in vitro*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3275 Supplier Abcam Supplier No. AB1446055MG
MW 465.8 Da, Purity >99%. Selective adrenergic-β3 receptor agonist. Induces adipose tissue remodelling in rat. Hypoglycemic agent. Active *in vivo* and *in vitro*.
The product is subject to the following: Abcam Restricted Use Statement