Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CL 316243 free acid 25mg | 183720-02-7 | 25 MG
SDP

Catalog No. 5000308025
Encompass_Preferred

CL 316243 free acid is a selective β3-adrenoceptor agonist provided as a white to off-white solid for research use. It is supplied with high reported purity and defined storage recommendations suitable for pharmacology studies involving β3-adrenergic receptor activity.

  • Selective β3-adrenoceptor agonist
  • High purity: 99.37%
  • Molecular weight 421.83 g/mol; formula C20H20ClNO7
  • White to off-white solid powder
  • Storage: powder -20°C (stable for 3 years); in solvent -80°C (6 months), -20°C (1 month)

Catalog No. 50-003-08025 Supplier Medchemexpress LLC Supplier No. HY11677125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

CL 316243 free acid is a selective β3-adrenoceptor agonist provided as a white to off-white solid for research use. It is supplied with high reported purity and defined storage recommendations suitable for pharmacology studies involving β3-adrenergic receptor activity.

  • Selective β3-adrenoceptor agonist
  • High purity: 99.37%
  • Molecular weight 421.83 g/mol; formula C20H20ClNO7
  • White to off-white solid powder
  • Storage: powder -20°C (stable for 3 years); in solvent -80°C (6 months), -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.