Learn More
Enzo Life Sciences CL-82198 (5mg). CAS: 307002-71-7
SDP
Catalog No. 502010685
Encompass_Preferred
A novel and selective inhibitor of MMP-13 (89% at 33 µM). Acts via binding to the S1' pocket of MMP-13 without metal chelation. Purity: ≥98% (TLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0685 Supplier Enzo Life Sciences Supplier No. BMLEI3020005
A novel and selective inhibitor of MMP-13 (89% at 33 µM). Acts via binding to the S1' pocket of MMP-13 without metal chelation. Purity: ≥98% (TLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: Ambient.