Learn More
Bellco Glass, Inc. CL A Graduated Cylinder,1000mL Double Metric Scale - TC
SDP
Catalog No. 501945731
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Used to contain and measure the volume of a liquid. Features hexagonal bumper guard and white double scale metric graduations. Size: 1l Dimensions: 70x422mm Graduation interval ml: 100 T
Catalog No. 50-194-5731 Supplier Bellco Glass, Inc. Supplier No. 562501000
Manufactured from Type 1 Class A Borosilicate Glass. Used to contain and measure the volume of a liquid. Features hexagonal bumper guard and white double scale metric graduations. Size: 1l Dimensions: 70x422mm Graduation interval ml: 100 T