Learn More
Bellco Glass, Inc. CL A Graduated Cylinder,100mL Double Metric Scale - TC
SDP
Catalog No. 501945728
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Used to contain and measure the volume of a liquid. Features hexagonal bumper guard and white double scale metric graduations. Size: 100ml Dimensions: 30x236mm Graduation interval ml: 0.10
Catalog No. 50-194-5728 Supplier Bellco Glass, Inc. Supplier No. 562500100
Manufactured from Type 1 Class A Borosilicate Glass. Used to contain and measure the volume of a liquid. Features hexagonal bumper guard and white double scale metric graduations. Size: 100ml Dimensions: 30x236mm Graduation interval ml: 0.10