Learn More
Bellco Glass, Inc. CL A Graduated Cylinder,2000mL Double Metric Scale - TC
SDP
Catalog No. 501945732
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Used to contain and measure the volume of a liquid. Features white double scale metric graduations. Size: 2l Dimensions: 90x478mm Graduation interval ml: 200 T
Catalog No. 50-194-5732 Supplier Bellco Glass, Inc. Supplier No. 562502000
Manufactured from Type 1 Class A Borosilicate Glass. Used to contain and measure the volume of a liquid. Features white double scale metric graduations. Size: 2l Dimensions: 90x478mm Graduation interval ml: 200 T