Learn More
Bellco Glass, Inc. CL A Graduated Cylinder,250mL Double Metric Scale - TC
SDP
Catalog No. 501945729
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Used to contain and measure the volume of a liquid. Features hexagonal bumper guard and white double scale metric graduations. Size: 250ml Dimensions: 41x301mm Graduation interval ml: 0.20
Catalog No. 50-194-5729 Supplier Bellco Glass, Inc. Supplier No. 562500250
Manufactured from Type 1 Class A Borosilicate Glass. Used to contain and measure the volume of a liquid. Features hexagonal bumper guard and white double scale metric graduations. Size: 250ml Dimensions: 41x301mm Graduation interval ml: 0.20