Learn More
Bellco Glass, Inc. CL A Graduated Cylinder,500mL Double Metric Scale - TC
SDP
Catalog No. 501945730
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Used to contain and measure the volume of a liquid. Features hexagonal bumper guard and white double scale metric graduations. Size: 500ml Dimensions: 53x362mm Graduation interval ml: 0.50
Catalog No. 50-194-5730 Supplier Bellco Glass, Inc. Supplier No. 562500500
Manufactured from Type 1 Class A Borosilicate Glass. Used to contain and measure the volume of a liquid. Features hexagonal bumper guard and white double scale metric graduations. Size: 500ml Dimensions: 53x362mm Graduation interval ml: 0.50