Learn More
Cayman Chemical Cl-Amidine (hydrochloride), 1373232-26-8, 10 mg
SDP
Supplier:  Cayman Chemical 1059910
Encompass_Preferred
Molecular Formula C14H19ClN4O2 • HCl, Purity ≥95%, Formula Weight 347.2, Synonyms: N-[(1S)-1-(aminocarbonyl)-4-[(2-chloro-1-iminoethyl)amino]butyl]-benzamide, monohydrochloride; A pan PAD inhibitor (IC50s = 0.8, 6.2, and 5.9 M for PAD1, PAD3, and PAD4, respectively, in vitro); irreversibly inactivates PAD4 (kinact/KI = 13,000 M-1min-1) and PAD3 (kinact/KI = 2,000 M-1min-1); cytotoxic to HL-60, MCF7, and HT-29 cancer cell lines (IC50s = 0.25, 0.05 and 1 M, respectively).
Catalog No. NC0393470