Learn More
Selleck Chemical LLC Cl-amidine 5mg
SDP
Supplier:  Selleck Chemical LLC S81415MG
Encompass_Preferred
Cl-amidine is an irreversible pan-peptidylarginine deiminase (PAD) inhibitor with IC50 values of 5.9 ¿ 0.3 ?M, 0.8 ¿ 0.3 ?M, 6.2 ¿ 1.0 ?M for PAD4, PAD1 and PAD3, respectively. Cl-amidine induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3343