Learn More
Medchemexpress LLC HY-100574A 10mg Medchemexpress, Cl-amidine (hydrochloride) CAS:1373232-26-8 Purity:>98%
SDP
Catalog No. 501931888
Encompass_Preferred
Medchemexpress, HY-100574A 10mg Cl-amidine (hydrochloride) CAS:1373232-26-8 Cl-amidine (hydrochloride) is a peptidylarginine deminase (PAD) inhibitor, with an IC 50 5.9 μM for PAD4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1888 Supplier Medchemexpress LLC Supplier No. HY100574A10MG
Medchemexpress, HY-100574A 10mg Cl-amidine (hydrochloride) CAS:1373232-26-8 Cl-amidine (hydrochloride) is a peptidylarginine deminase (PAD) inhibitor, with an IC 50 5.9 μM for PAD4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.