Learn More
Medchemexpress LLC Cl-amidine (hydrochloride) | 1373232-26-8 | 99.8% | 347.24 | C14H20Cl2N4O2 | 5 MG
SDP
Catalog No. 5000212989
Encompass_Preferred
Cl-amidine (hydrochloride) is a high-purity small-molecule inhibitor of peptidylarginine deiminases (PADs), used in biochemical and cellular research to block protein citrullination and probe PAD-mediated pathways.
  • Potent inhibition of PAD enzymes, including PAD1, PAD3, and PAD4.
  • High chemical purity suitable for biochemical assays.
  • Solid form enables flexible preparation of solutions.
  • Stable when stored at recommended low temperatures.
  • Available in milligram quantities for research use.
Catalog No. 50-002-12989 Supplier Medchemexpress LLC Supplier No. HY100574A5MG
Cl-amidine (hydrochloride) is a high-purity small-molecule inhibitor of peptidylarginine deiminases (PADs), used in biochemical and cellular research to block protein citrullination and probe PAD-mediated pathways.
  • Potent inhibition of PAD enzymes, including PAD1, PAD3, and PAD4.
  • High chemical purity suitable for biochemical assays.
  • Solid form enables flexible preparation of solutions.
  • Stable when stored at recommended low temperatures.
  • Available in milligram quantities for research use.