Learn More
Medchemexpress LLC Cl-amidine hydrochloride | 1373232-26-8 | MFCD22201084 | 99.8% | 347.24 | C14H20Cl2N4O2 | 1 ML
SDP
Catalog No. 5000211540
Encompass_Preferred
Cl-amidine hydrochloride is an orally active peptidylarginine deiminase (PAD) inhibitor used in biochemical, cellular, and in vivo research to block protein citrullination and probe PAD-related pathways.
  • Inhibits PAD1, PAD3, and PAD4 with reported IC50 values of 0.8 μM, 6.2 μM, and 5.9 μM.
  • Available as a 10 mM solution in DMSO or as solid quantities for formulation flexibility.
  • High purity (~99.8%) suitable for biochemical and cellular assays.
  • Demonstrated activity in cell apoptosis and neutrophil extracellular trap formation assays.
  • Shows efficacy in murine models of colitis and sepsis in published studies.
  • Provided supporting documents: data sheet, SDS, COA, HNMR, RP-HPLC, and LCMS.
Catalog No. 50-002-11540 Supplier Medchemexpress LLC Supplier No. HY100574A10MM 1ML
Cl-amidine hydrochloride is an orally active peptidylarginine deiminase (PAD) inhibitor used in biochemical, cellular, and in vivo research to block protein citrullination and probe PAD-related pathways.
  • Inhibits PAD1, PAD3, and PAD4 with reported IC50 values of 0.8 μM, 6.2 μM, and 5.9 μM.
  • Available as a 10 mM solution in DMSO or as solid quantities for formulation flexibility.
  • High purity (~99.8%) suitable for biochemical and cellular assays.
  • Demonstrated activity in cell apoptosis and neutrophil extracellular trap formation assays.
  • Shows efficacy in murine models of colitis and sepsis in published studies.
  • Provided supporting documents: data sheet, SDS, COA, HNMR, RP-HPLC, and LCMS.