Learn More
Medchemexpress LLC Cl-amidine hydrochloride | 1373232-26-8 | MFCD22201084 | 99.8% | 347.24 g·mol⁻¹ | C14H20Cl2N4O2 | 1 MG
SDP
Catalog No. 5000212991
Encompass_Preferred
Cl-amidine hydrochloride is an orally active peptidylarginine deiminase (PAD) inhibitor used in biochemical and cellular research to probe histone citrullination, neutrophil extracellular trap formation, apoptosis, and related signaling pathways. Reported IC50 values are ~0.8 μM (PAD1), 6.2 μM (PAD3), and 5.9 μM (PAD4). Supplied as a high-purity solid; follow manufacturer storage recommendations.
  • Potent inhibitor of PAD enzymes with reported low-micromolar activity.
  • High purity suitable for biochemical assays and cell-based studies.
  • Stable solid formulation; store under inert atmosphere at low temperature.
  • Available in solution to facilitate cell treatment workflows.
  • For research use only; not for human or veterinary applications.
Catalog No. 50-002-12991 Supplier Medchemexpress LLC Supplier No. HY100574A1MG
Cl-amidine hydrochloride is an orally active peptidylarginine deiminase (PAD) inhibitor used in biochemical and cellular research to probe histone citrullination, neutrophil extracellular trap formation, apoptosis, and related signaling pathways. Reported IC50 values are ~0.8 μM (PAD1), 6.2 μM (PAD3), and 5.9 μM (PAD4). Supplied as a high-purity solid; follow manufacturer storage recommendations.
  • Potent inhibitor of PAD enzymes with reported low-micromolar activity.
  • High purity suitable for biochemical assays and cell-based studies.
  • Stable solid formulation; store under inert atmosphere at low temperature.
  • Available in solution to facilitate cell treatment workflows.
  • For research use only; not for human or veterinary applications.