Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cl-C6-PEG4-C3-COOH | 2127391-58-4 | MFCD34469791 | 99.7% | 382.92 | C18H35ClO6 | 5 MG
SDP

Catalog No. 5000222885
Encompass_Preferred

Cl-C6-PEG4-C3-COOH is a bifunctional PROTAC linker that incorporates a chloroalkane handle for the synthesis of HaloPROTACs and other chloroalkane-containing bifunctional molecules. It is supplied as a high-purity liquid and is intended as a synthetic building block for chemical biology and drug discovery research.

  • Bifunctional PROTAC linker for constructing chloroalkane-containing PROTACs.
  • Molecular weight 382.92.
  • CAS number 2127391-58-4.
  • Purity 99.65%.
  • Physical form: colorless to light yellow liquid; density 1.072 g/cm3.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Storage: store at 4°C, protect from light; in solvent store -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-22885 Supplier Medchemexpress LLC Supplier No. HY1353065MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cl-C6-PEG4-C3-COOH is a bifunctional PROTAC linker that incorporates a chloroalkane handle for the synthesis of HaloPROTACs and other chloroalkane-containing bifunctional molecules. It is supplied as a high-purity liquid and is intended as a synthetic building block for chemical biology and drug discovery research.

  • Bifunctional PROTAC linker for constructing chloroalkane-containing PROTACs.
  • Molecular weight 382.92.
  • CAS number 2127391-58-4.
  • Purity 99.65%.
  • Physical form: colorless to light yellow liquid; density 1.072 g/cm3.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Storage: store at 4°C, protect from light; in solvent store -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.