Learn More
Medchemexpress LLC HY-112496 100mg Medchemexpress, Cl-C6-PEG4-O-CH2COOH CAS:1799506-30-1 Purity:>98%
SDP
Catalog No. 502080817
Encompass_Preferred
Medchemexpress, HY-112496 100mg Cl-C6-PEG4-O-CH2COOH CAS:1799506-30-1 Cl-C6-PEG4-O-CH2COOH (PROTAC Linker 4) is a PEG-based PROTAC linker can be used in the synthesis of chloroalkane-containing PROTACs (HaloPROTACs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0817 Supplier Medchemexpress LLC Supplier No. HY112496100MG
Medchemexpress, HY-112496 100mg Cl-C6-PEG4-O-CH2COOH CAS:1799506-30-1 Cl-C6-PEG4-O-CH2COOH (PROTAC Linker 4) is a PEG-based PROTAC linker can be used in the synthesis of chloroalkane-containing PROTACs (HaloPROTACs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.