Learn More
Medchemexpress LLC Cl-C6-PEG4-O-CH2COOH | 1799506-30-1 | 98.0% | 370.87 | 1 G
SDP
Catalog No. 5000436509
Encompass_Preferred
Cl-C6-PEG4-O-CH2COOH is a PEG-based PROTAC linker designed for the synthesis of chloroalkane-containing PROTACs (HaloPROTACs). This linker facilitates the formation of PROTACs, which are molecules that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker.
  • Used in the synthesis of chloroalkane-containing PROTACs (HaloPROTACs).
  • Facilitates PROTACs with two ligands for E3 ubiquitin ligase and target protein.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
Catalog No. 50-004-36509 Supplier Medchemexpress LLC Supplier No. HY1124961G
Cl-C6-PEG4-O-CH2COOH is a PEG-based PROTAC linker designed for the synthesis of chloroalkane-containing PROTACs (HaloPROTACs). This linker facilitates the formation of PROTACs, which are molecules that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker.
  • Used in the synthesis of chloroalkane-containing PROTACs (HaloPROTACs).
  • Facilitates PROTACs with two ligands for E3 ubiquitin ligase and target protein.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.