Learn More
TARGETMOL CHEMICALS INC CL-C6-PEG4-O-CH2COOH 50MG
SDP
Catalog No. 502581072
Encompass_Preferred
Also available in 5 mg 10 mg 100 mg and bulk. Please contact Fisher for quotes. Cl-C6-PEG4-O-CH2COOH (PROTAC Linker 4) is an effective pegylated PROTAC linker which is often used in PROTAC synthesis of chloroanes (HaloPROTACs). purity: 99%
Catalog No. 50-258-1072 Supplier TARGETMOL CHEMICALS INC Supplier No. T1863950MG
Also available in 5 mg 10 mg 100 mg and bulk. Please contact Fisher for quotes. Cl-C6-PEG4-O-CH2COOH (PROTAC Linker 4) is an effective pegylated PROTAC linker which is often used in PROTAC synthesis of chloroanes (HaloPROTACs). purity: 99%