Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

InBios International Inc. CL Detect™ Rapid Test for Cutaneous Leishmaniasis
SDP

Catalog No. 21931260
Change view
Click to view available options
Quantity:
25 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-931-260 25 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-931-260 Supplier InBios International Inc. Supplier No. CL025
Add to Cart
Add to Cart
This item is not returnable. View return policy

The CL Detect™ Rapid Test is a qualitative, in vitro immunochromatographic assay for the rapid detection of Leishmania species antigen in ulcerative skin lesions.

  • FDA 510(k) Cleared and CE Marked
  • Relative Specificity in non-endemic population: 96%
  • Relative Sensitivity and Specificity in endemic population: 100% and 84% respectively
  • Kit includes 25 tests, controls, buffers and sterile dental broaches

Specifications

Content And Storage Room Temp, 20°C to 30°C
Detectable Analytes Leishmania species antigen
Detection Method Rapid Test
Type Rapid Test
Immunoassay Kit Format ICT (Strip)
Quantity 25 Tests/Kit
Analysis Time 20 min.
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.