Learn More
Bioss Cl. Difficile
SDP
Catalog No. 5000066344
Encompass_Preferred
Cl Difficile Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-66344 Supplier Bioss Supplier No. BSM72065M
Cl Difficile Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide