Learn More
Medchemexpress LLC CL-L1/COLEC10 Mouse 2ug | 2ug
SDP
Catalog No. 5000185806
Encompass_Preferred
CL-L1/COLEC10 operating as a lectin displays distinct sugar-binding preferences with a hierarchy of galactose mannose fucose N-acetylglucosamine N-acetylgalactosamine Apart from its sugar-binding role CL-L1/COLEC10 functions as a chemoattractant suggesting potential engagement in the modulation of cell migration CL-L1/COLEC10 Protein Mouse (HEK293 Fc) is the recombinant mouse-derived CL-L1/COLEC10 protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-85806 Supplier Medchemexpress LLC Supplier No. HYP768452UG
CL-L1/COLEC10 operating as a lectin displays distinct sugar-binding preferences with a hierarchy of galactose mannose fucose N-acetylglucosamine N-acetylgalactosamine Apart from its sugar-binding role CL-L1/COLEC10 functions as a chemoattractant suggesting potential engagement in the modulation of cell migration CL-L1/COLEC10 Protein Mouse (HEK293 Fc) is the recombinant mouse-derived CL-L1/COLEC10 protein expressed by HEK293 with N-hFc labeled tag