Learn More
Cayman Chemical Cl-ncrostatIn-1 10mg
SDP
Catalog No. 502952065
Encompass_Preferred
A RIPK1 inhibitor; inhibits RIPK1 activity at 0.3, 3, and 30 µM; inhibits TNF-α-induced necroptosis in FADD-deficient Jurkat cells (EC50 = 180 nM); reduces infarct size when administered i.c.v. pre- and post-occlusion, or post-occlusion only, as 2 µl of a 4 mM solution in a mouse model of MCAO
Catalog No. 50-295-2065 Supplier Cayman Chemical Supplier No. 1165610 MG
A RIPK1 inhibitor; inhibits RIPK1 activity at 0.3, 3, and 30 µM; inhibits TNF-α-induced necroptosis in FADD-deficient Jurkat cells (EC50 = 180 nM); reduces infarct size when administered i.c.v. pre- and post-occlusion, or post-occlusion only, as 2 µl of a 4 mM solution in a mouse model of MCAO