Learn More
Medchemexpress LLC Cl-NQTrp | 185351-23-9 | MFCD34471035 | 100.0% | 394.8 g·mol⁻¹ | C21H15ClN2O4 | 5 MG
SDP
Catalog No. 5000221399
Encompass_Preferred
Cl-NQTrp is a small-molecule research reagent reported to disrupt preformed fibrillar aggregates of the Tau-derived PHF6 (VQIVYK) peptide and full-length tau protein. Supplied as a high-purity solid, it is intended for in vitro biochemical and cellular studies of tau aggregation and disaggregation.
  • Disrupts preformed tau and PHF6 fibrillar aggregates.
  • Suitable for in vitro biochemical and cellular assays.
  • High purity (99.97%) for reproducible results.
  • Soluble in DMSO for stock solution preparation.
  • Available in small research pack sizes for screening studies.
Catalog No. 50-002-21399 Supplier Medchemexpress LLC Supplier No. HY1386435MG
Cl-NQTrp is a small-molecule research reagent reported to disrupt preformed fibrillar aggregates of the Tau-derived PHF6 (VQIVYK) peptide and full-length tau protein. Supplied as a high-purity solid, it is intended for in vitro biochemical and cellular studies of tau aggregation and disaggregation.
  • Disrupts preformed tau and PHF6 fibrillar aggregates.
  • Suitable for in vitro biochemical and cellular assays.
  • High purity (99.97%) for reproducible results.
  • Soluble in DMSO for stock solution preparation.
  • Available in small research pack sizes for screening studies.