Learn More
Medchemexpress LLC Cl-PEG2-acid | 170304-76-4 | 182.60 | 250 MG
SDP
Catalog No. 5000402738
Encompass_Preferred
Cl-PEG2-acid is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Facilitates selective degradation of target proteins
  • Utilizes the ubiquitin-proteasome system
Catalog No. 50-004-02738 Supplier Medchemexpress LLC Supplier No. 250MGHY43055
Cl-PEG2-acid is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Facilitates selective degradation of target proteins
  • Utilizes the ubiquitin-proteasome system