Learn More
Medchemexpress LLC Cl-PEG4-acid 50mg
SDP
Catalog No. 5000205795
Encompass_Preferred
Cl-PEG4-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-05795 Supplier Medchemexpress LLC Supplier No. HYW09612950MG
Cl-PEG4-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]