Learn More
Foxx Life Sciences Abdos Vacuum Desiccator with a clear Polycarbonate (PC) top and Plate (Diameter 200mm) 1/EA
GREENER_CHOICE
Catalog No. NC1818253
Encompass_Preferred
These All Clear Polycarbonate (PC) Vacuum desiccators are molded with high-quality plastics which are light and easy to handle. Desiccators are enclosures containing desiccants for preserving moisture sensitive products and the vacuum chamber accelerates drying. Silicone O-ring assures tight seal down to 0°C without using grease. Bottom part of the desiccators comes with thick perforated polypropylene disc with big holes in the middle for easy lifting.•Polypropylene stopcock with PTFEFoxx Life Sciences is proud to represent high-quality Abdos plasticware and plastic laboratory products. For downloadable Certificates please visit: http://www.abdoslifesciences.com/Global/certificates.html.
Catalog No. NC1818253 Supplier Foxx Life Sciences Supplier No. E11610
These All Clear Polycarbonate (PC) Vacuum desiccators are molded with high-quality plastics which are light and easy to handle. Desiccators are enclosures containing desiccants for preserving moisture sensitive products and the vacuum chamber accelerates drying. Silicone O-ring assures tight seal down to 0°C without using grease. Bottom part of the desiccators comes with thick perforated polypropylene disc with big holes in the middle for easy lifting.•Polypropylene stopcock with PTFEFoxx Life Sciences is proud to represent high-quality Abdos plasticware and plastic laboratory products. For downloadable Certificates please visit: http://www.abdoslifesciences.com/Global/certificates.html.