Learn More
Hach Company Chlorine Dioxide Reagent Set, DPD Glycine, 10 mL
Supplier:  Hach Company 2770900
Encompass
For determination of Chiorine Dioxide by the DPD Glycine method. USEPA accepted for reporting for drinking water analysis using Hach Method 10126. Range: 0.04 - 5.00 mg/L ClO₂. Sample Size: 10 mL Reagent set includes DPD Free Chlorine Reagent Powder Pillows and Glycine Reagent for approximately 100 tests.Boric Acid free!
Catalog No. 50-199-6661