Learn More
Invivogen CL097, 5 mg
SDP
Catalog No. NC1203867
Encompass
CL097 is a highly water-soluble derivative of the imidazoquinoline compound R848 (Resiquimod). Like R848, CL097 induces TLR7 and/or TLR8 responses in human and murine immune cells.
Catalog No. NC1203867 Supplier Invivogen Supplier No. TLRLC975
CL097 is a highly water-soluble derivative of the imidazoquinoline compound R848 (Resiquimod). Like R848, CL097 induces TLR7 and/or TLR8 responses in human and murine immune cells.